missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nalpha-Boc-N^w-(2,2,4,6,7-pentamethyl-2,3-dihydrobenzo[b]furan-5-sulfonyl)-L-arginine, 96%, Thermo Scientific Chemicals
£42.50 - £158.00
Chemical Identifiers
CAS | 200124-22-7 |
---|---|
Molecular Formula | C24H38N4O7S |
Molecular Weight (g/mol) | 526.649 |
MDL Number | MFCD00236821 |
InChI Key | CVFXPOKENLGCID-KRWDZBQOSA-N |
Synonym | boc-arg pbf-oh, s-2-tert-butoxycarbonyl amino-5-3-2,2,4,6,7-pentamethyl-2,3-dihydrobenzofuran-5-yl sulfonyl guanidino pentanoic acid, n-boc-n'-2,2,4,6,7-pentamethyldihydrobenzofuran-5-sufonyl-l-arginine, ambotzbaa1067, boc-arg pbf-0h, n,a-boc-n, o-pbf-l-arginine, boc-arg pbf-oh tlc, o-2,2,4,6,7-pentamethyl-dihydrobenzofuran-5-sulfonyl-l-arginine, 2s-2-tert-butoxycarbonyl amino-5-n'-2,2,4,6,7-pentamethyl-3h-1-benzofuran-5-ylsulfonyl carbamimidamido pentanoic acid, 2s-5-e-amino 2,2,4,6,7-pentamethyl-3h-1-benzofuran-5-sulfonamido methylidene amino-2-tert-butoxycarbonyl amino pentanoic acid |
PubChem CID | 11341472 |
IUPAC Name | (2S)-5-[[amino-[(2,2,4,6,7-pentamethyl-3H-1-benzofuran-5-yl)sulfonylamino]methylidene]amino]-2-[(2-methylpropan-2-yl)oxycarbonylamino]pentanoic acid |
SMILES | CC1=C2C(=C(C(=C1C)S(=O)(=O)NC(=NCCCC(C(=O)O)NC(=O)OC(C)(C)C)N)C)CC(O2)(C)C |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15435545
|
Thermo Scientific Alfa Aesar
H62148.03 |
1 g |
£42.50
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15445545
|
Thermo Scientific Alfa Aesar
H62148.06 |
5 g |
£158.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Nalpha-Boc-Nμ-(2,2,4,6,7-pentamethyl-2,3-dihydrobenzo[b]furan-5-sulfonyl)-L-arginine is used as a triflating reagent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsNalpha-Boc-Nµ-(2,2,4,6,7-pentamethyl-2,3-dihydrobenzo[b]furan-5-sulfonyl)-L-arginine is used as a triflating reagent.
Solubility
Soluble in water or 1% acetic acid
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Chemical Identifiers
200124-22-7 | |
526.649 | |
CVFXPOKENLGCID-KRWDZBQOSA-N | |
11341472 | |
CC1=C2C(=C(C(=C1C)S(=O)(=O)NC(=NCCCC(C(=O)O)NC(=O)OC(C)(C)C)N)C)CC(O2)(C)C |
C24H38N4O7S | |
MFCD00236821 | |
boc-arg pbf-oh, s-2-tert-butoxycarbonyl amino-5-3-2,2,4,6,7-pentamethyl-2,3-dihydrobenzofuran-5-yl sulfonyl guanidino pentanoic acid, n-boc-n'-2,2,4,6,7-pentamethyldihydrobenzofuran-5-sufonyl-l-arginine, ambotzbaa1067, boc-arg pbf-0h, n,a-boc-n, o-pbf-l-arginine, boc-arg pbf-oh tlc, o-2,2,4,6,7-pentamethyl-dihydrobenzofuran-5-sulfonyl-l-arginine, 2s-2-tert-butoxycarbonyl amino-5-n'-2,2,4,6,7-pentamethyl-3h-1-benzofuran-5-ylsulfonyl carbamimidamido pentanoic acid, 2s-5-e-amino 2,2,4,6,7-pentamethyl-3h-1-benzofuran-5-sulfonamido methylidene amino-2-tert-butoxycarbonyl amino pentanoic acid | |
(2S)-5-[[amino-[(2,2,4,6,7-pentamethyl-3H-1-benzofuran-5-yl)sulfonylamino]methylidene]amino]-2-[(2-methylpropan-2-yl)oxycarbonylamino]pentanoic acid |
Specifications
200124-22-7 | |
MFCD00236821 | |
Soluble in water or 1% acetic acid | |
CC1=C2C(=C(C(=C1C)S(=O)(=O)NC(=NCCCC(C(=O)O)NC(=O)OC(C)(C)C)N)C)CC(O2)(C)C | |
526.649 | |
526.65 | |
Nalpha-Boc-Nomega-(2,2,4,6,7-pentamethyl-2,3-dihydrobenzo[b]furan-5-sulfonyl)-L-arginine |
C24H38N4O7S | |
boc-arg pbf-oh, s-2-tert-butoxycarbonyl amino-5-3-2,2,4,6,7-pentamethyl-2,3-dihydrobenzofuran-5-yl sulfonyl guanidino pentanoic acid, n-boc-n'-2,2,4,6,7-pentamethyldihydrobenzofuran-5-sufonyl-l-arginine, ambotzbaa1067, boc-arg pbf-0h, n,a-boc-n, o-pbf-l-arginine, boc-arg pbf-oh tlc, o-2,2,4,6,7-pentamethyl-dihydrobenzofuran-5-sulfonyl-l-arginine, 2s-2-tert-butoxycarbonyl amino-5-n'-2,2,4,6,7-pentamethyl-3h-1-benzofuran-5-ylsulfonyl carbamimidamido pentanoic acid, 2s-5-e-amino 2,2,4,6,7-pentamethyl-3h-1-benzofuran-5-sulfonamido methylidene amino-2-tert-butoxycarbonyl amino pentanoic acid | |
CVFXPOKENLGCID-KRWDZBQOSA-N | |
(2S)-5-[[amino-[(2,2,4,6,7-pentamethyl-3H-1-benzofuran-5-yl)sulfonylamino]methylidene]amino]-2-[(2-methylpropan-2-yl)oxycarbonylamino]pentanoic acid | |
11341472 | |
96% |
Safety and Handling
missing translation for 'tsca' : No
Recommended Storage : Keep cold
RUO – Research Use Only