missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nalpha-Boc-Nepsilon-trifluoroacetyl-L-lysine, 95%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
25 g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
N alpha-Boc-N epsilon-trifluoroacetyl-L-lysine, is used as a fine chemical intermediate.
Solubility
Soluble in DMSO.
Notes
Heat sensitive. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
N alpha-Boc-N epsilon-trifluoroacetyl-L-lysine, is used as a fine chemical intermediate.
Solubility
Soluble in DMSO.
Notes
Heat sensitive. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
Specifications
Specifications
Color | White |
Solubility Information | Soluble in DMSO. |
Formula Weight | 342.31 |
Percent Purity | 95% |
Quantity | 1 g |
Physical Form | Powder |
Chemical Name or Material | Nalpha-Boc-Nepsilon-trifluoroacetyl-L-lysine |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'