missing translation for 'onlineSavingsMsg'
Learn More

Nalpha-Boc-Ngamma-trityl-L-asparagine, 98%, Thermo Scientific Chemicals

Product Code. 15448958
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 132388-68-2
Molecular Formula C28H30N2O5
Molecular Weight (g/mol) 474.557
MDL Number MFCD00153299
InChI Key PYGOCFDOBSXROC-QHCPKHFHSA-N
Synonym boc-asn trt-oh, nalpha-boc-ngamma-trityl-l-asparagine, nalpha-tert-butoxycarbonyl-ngamma-trityl-l-asparagine, s-2-tert-butoxycarbonyl amino-4-oxo-4-tritylamino butanoic acid, boc-n-beta-trityl-l-asparagine, 2s-2-tert-butoxycarbonyl amino-3-triphenylmethylcarbamoyl propanoic acid, boc-ngamma-trityl-d-asparagine, boc-asn-trt-oh, pubchem18974, n, a-boc-n, a-trityl-l-asparagine
PubChem CID 11385960
IUPAC Name (2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-4-oxo-4-(tritylamino)butanoic acid
SMILES CC(C)(C)OC(=O)NC(CC(=O)NC(C1=CC=CC=C1)(C2=CC=CC=C2)C3=CC=CC=C3)C(=O)O

Specifications

Melting Point 202°C to 205°C
Quantity 25 g
Beilstein 4339761
Formula Weight 474.55
Percent Purity 98%
Chemical Name or Material Nalpha-Boc-Ngamma-trityl-L-asparagine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'