missing translation for 'onlineSavingsMsg'
Learn More

Nalpha-Fmoc-N^d-trityl-D-glutamine, 98%, Thermo Scientific Chemicals

Product Code. 15449058
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 200623-62-7
Molecular Formula C39H34N2O5
Molecular Weight (g/mol) 610.71
MDL Number MFCD00151924
InChI Key WDGICUODAOGOMO-PGUFJCEWSA-N
Synonym fmoc-d-gln trt-oh, n-fmoc-n'-trityl-d-glutamine, fmoc-gln trt, 2r-2-9h-fluoren-9-ylmethoxy carbonyl amino-4-triphenylmethylcarbamoyl butanoic acid, ambotzfaa1322, pubchem12399, n-alpha-9-fluorenylmethyloxycarbonyl-n-gamma-trityl-d-glutamine, n∼2∼-9h-fluoren-9-yl methoxy carbonyl-n-triphenylmethyl-d-glutamine, 2r-2-9h-fluoren-9-yl methoxy carbonyl amino-4-triphenylmethyl carbamoyl butanoic acid
PubChem CID 24820181
IUPAC Name (2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-5-oxo-5-(tritylamino)pentanoic acid
SMILES C1=CC=C(C=C1)C(C2=CC=CC=C2)(C3=CC=CC=C3)NC(=O)CCC(C(=O)O)NC(=O)OCC4C5=CC=CC=C5C6=CC=CC=C46

Specifications

Quantity 1 g
Formula Weight 610.7
Percent Purity 98%
Chemical Name or Material Nalpha-Fmoc-Ndelta-trityl-D-glutamine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'