missing translation for 'onlineSavingsMsg'
Learn More

Nalpha-Fmoc-Ndelta-trityl-L-glutamine pentafluorophenyl ester, 97%, Thermo Scientific™

Product Code. 11436340
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 132388-65-9
Molecular Formula C45H33F5N2O5
Molecular Weight (g/mol) 776.76
MDL Number MFCD00800874
InChI Key OLYKAAWLZTVISS-DHUJRADRSA-N
Synonym fmoc-gln trt-opfp, fmoc-l-gln trt-opfp, fmoc-glutamine trt-opfp, fmoc-n-delta-trityl-l-glutamine pentafluorophenyl ester, fmoc-n-gamma-trityl-l-glutamine pentafluorophenyl ester, n-fmoc-n'-trityl-l-glutamine pentafluorophenyl ester, n, a-fmoc-n, a-trityl-l-glutamine pentafluorophenyl ester, n-alpha-9-fluorenylmethyloxycarbonyl-n-gamma-tri, n-alpha-fmoc-gamma-trityl-l-glutamine pentafluorophenyl ester
PubChem CID 11297250
IUPAC Name (2,3,4,5,6-pentafluorophenyl) (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-5-oxo-5-(tritylamino)pentanoate
SMILES C1=CC=C(C=C1)C(C2=CC=CC=C2)(C3=CC=CC=C3)NC(=O)CCC(C(=O)OC4=C(C(=C(C(=C4F)F)F)F)F)NC(=O)OCC5C6=CC=CC=C6C7=CC=CC=C57

Specifications

Melting Point 183°C to 184°C
Quantity 5 g
Optical Rotation −16° (c=1 in THF)
Formula Weight 776.75
Percent Purity 97%
Chemical Name or Material Nalpha-Fmoc-Ndelta-trityl-L-glutamine pentafluorophenyl ester
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'