Learn More
Nano-Secondary™ Alpaca, anti-Human, CoraLite Plus 488, Chromotek
Alpaca Recombinant Monoclonal Antibody
Brand:  Chromotek shuGCL488-2-100
 View more versions of this product
Product Code. 18015475
  • £276.00 / 100µL
Explore more special offers
Description
Description
Nano-Secondary™ alpaca anti-human IgG, recombinant VHH, is a human IgG-specific (IgG1, IgG2, IgG3, IgG4) antibody. This secondary antibody product consists of a Nanobody that binds to the Fc-domain of human IgG with high affinity and specificity.
Specifications
Specifications
Nano-Secondary™
Alpaca
Secondary
Liquid
CoraLite Plus 488
100 μL
Human
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Nano-Secondary™ Alpaca, anti-Human, CoraLite Plus 488, Chromotek > Quantity: 100 μL