missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nano-Secondary™ Alpaca, anti-Human, CoraLite Plus 647, Chromotek
Alpaca Recombinant Monoclonal Antibody
£76.00 - £276.00
Specifications
Antigen | Nano-Secondary™ |
---|---|
Conjugate | CoraLite Plus 647 |
Form | Liquid |
Host Species | Alpaca |
Target Species | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18006865
|
Chromotek
shuGCL647-2-10 |
10 μL |
£76.00
10µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18058695
|
Chromotek
shuGCL647-2-100 |
100 μL |
£276.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Nano-Secondary™ alpaca anti-human IgG, recombinant VHH, is a human IgG-specific (IgG1, IgG2, IgG3, IgG4) antibody. This secondary antibody product consists of a Nanobody that binds to the Fc-domain of human IgG with high affinity and specificity.
Specifications
Nano-Secondary™ | |
Liquid | |
Human | |
Secondary |
CoraLite Plus 647 | |
Alpaca | |
IgG |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Nano-Secondary™ Alpaca, anti-Human, CoraLite Plus 647, Chromotek