Learn More
Nano-Secondary™ Alpaca, anti-Human, CoraLite Plus 750, Chromotek
Alpaca Recombinant Monoclonal Antibody
Brand: Chromotek shuGCL750-2-10
Additional Details : Weight : 0.00100kg
Description
Nano-Secondary™ alpaca anti-human IgG, recombinant VHH, is a human IgG-specific (IgG1, IgG2, IgG3, IgG4) antibody. This secondary antibody product consists of a Nanobody that binds to the Fc-domain of human IgG with high affinity and specificity.
Specifications
Nano-Secondary™ | |
Alpaca | |
Secondary | |
Liquid |
CoraLite Plus 750 | |
10 μL | |
Human | |
IgG |