missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific NanoDrop™ Reagents

Product Code. p-10341000 Shop All Thermo Scientific Products
Click to view available options
Type:
PV-8 Performance Verification
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 30089480

Brand: Thermo Scientific CHEMPV8

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Certified standards for verifying instrument performance to factory specifications are available separately.

For use with NanoDrop Eight UV-Vis Spectrophotometer Solution with certified absorbance values. Use to create a record of correct photometric performance.

Contains: Two vials of CHEM-PV-1, two pastuer pipettes, and two 8-well PCR strip tubes.

Intended for single use only.

TRUSTED_SUSTAINABILITY

Specifications

Type PV-8 Performance Verification
For Use With (Equipment) NanoDrop Eight
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.