missing translation for 'onlineSavingsMsg'
Learn More

Biochrom NanoVue Plus Micro-Volume UV-Vis Spectrophotometer with Printer

Easy to use instrument designed for reliable measurements of nucleic acids and proteins. Includes printer and operates without the use of a PC.

Brand:  Biochrom 28956058


Code : GE

Additional Details : Weight : 6.00900kg

Product Code. 10794467

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

  • IQ/OQ Documents available
  • Gold-colored sample plate enhances compatibility with the wide range of chemicals and sample types used in today's busy laboratories.
  • Predefined methods for protein quantitation, including direct UV, Bradford, BCA, Biuret, and Lowry, with capability to run up to 27 standards.
  • Accurately assays low volumes of 0.5 to 5μL conserving precious samples.
  • Two pathlengths - 0.2mm and 0.5mm
  • Convenient, drop and read sample plate eliminates the need for cuvettes and capillaries.
  • Predefined methods for nucleic acid quantitation, including concentration and purity.
  • UV-Visible spectrophotometer for quick and accurate quantitation of nucleic acids and protein samples.
  • No PC: Large, high-resolution graphical display enables a quick read of relevant results.
  • Calibration curves, kinetics or ratio measurements are all displayed at the touch of a button.
  • Hydrophobic plate reduces sample carry over and allows for excellent sample recovery.
Specifications

Specifications

Max. ±1% at 259 nm (at 0.7 to 0.8 A using uracil)
260 x 390 x 100 mm
5 nm
200 to 1100 nm
50/60 Hz
Included
±0.5 nm
Twin CCD Array
Xenon
Nanoplus with Printer
100/240 V ±10%, 50/60 Hz
0.2 mm or 0.5 mm (automatic or manual)
±2 nm
4.5 kg
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers