missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva (formerly Pall Lab) Nanosep™ MF Centrifugal Devices with Bio-Inert™ Membrane
Use Nanosep™ devices to concentrate, purify, and desalt peptides, proteins, oligonucleotides, DNA, and RNA.
£108.00 - £1460.00
Specifications
Description | EFA: 0.3 cm2 |
---|---|
Material (Membrane) | Bio-Inert (modified nylon) |
Sterility | Non-sterile |
Housing Material | Polypropylene |
Volume (Metric) | 50 to 500 μL (Sample) |
Product Code | Brand | Pore Size | Includes | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Pore Size | Includes | Price | Quantity & Availability | ||||
17124801
|
Cytiva (formerly Pall Lab)
ODM45C33 |
0.45 μm | 24/Pk. |
£108.00
Pack of 24 |
Estimated Shipment: 30-08-2024
Log in to see stock. |
||||
17104801
|
Cytiva (formerly Pall Lab)
ODM02C34 |
0.2 μm | 100/Pk. |
£358.00
Pack of 100 |
Estimated Shipment: 07-08-2024
Log in to see stock. |
||||
17194791
|
Cytiva (formerly Pall Lab)
ODM02C33 |
0.2 μm | 24/Pk. |
£138.00
Pack of 24 |
Estimated Shipment: 19-11-2024
Log in to see stock. |
||||
17114801
|
Cytiva (formerly Pall Lab)
ODM02C35 |
0.2 μm | 500/Pk. |
£1460.00
Pack of 500 |
Estimated Shipment: 07-08-2024
Log in to see stock. |
||||
17134801
|
Cytiva (formerly Pall Lab)
ODM45C34 |
0.45 μm | 100/Pk. |
£401.00
Pack of 100 |
Estimated Shipment: 30-08-2024
Log in to see stock. |
||||
17144801
|
Cytiva (formerly Pall Lab)
ODM45C35 |
0.45 μm | 500/Pk. |
£1460.00
Pack of 500 |
Estimated Shipment: 30-08-2024
Log in to see stock. |
||||
Description
- Simple, reliable,concentrating and desalting of 50 to 500 μL samples
- This product is not approved for use in medical, clinical, surgical or other patient applications
Specifications
EFA: 0.3 cm2 | |
Non-sterile | |
50 to 500 μL (Sample) |
Bio-Inert (modified nylon) | |
Polypropylene |