missing translation for 'onlineSavingsMsg'
Learn More

Naphthalene-1,4,5,8-tetracarboxylic acid dianhydride, 97%, Thermo Scientific Chemicals

Product Code. 11383496
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Naphthalene-1,4,5,8-tetracarboxylic acid dianhydride is used as a reagent in the preparation of novel fluorescent markers for hypoxic cells of naphthalimides with two heterocyclic side chains for bioreductive binding.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Naphthalene-1,4,5,8-tetracarboxylic acid dianhydride is used as a reagent in the preparation of novel fluorescent markers for hypoxic cells of naphthalimides with two heterocyclic side chains for bioreductive binding.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. It is sensitive to moisture. Store under inert gas. Incompatible with acids, bases and oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 81-30-1
Molecular Formula C14H4O6
Molecular Weight (g/mol) 268.18
MDL Number MFCD00006915
InChI Key YTVNOVQHSGMMOV-UHFFFAOYSA-N
Synonym 1,4,5,8-naphthalenetetracarboxylic dianhydride, ntcda, naphthalenetetracarboxylic dianhydride, isochromeno 6,5,4-def isochromene-1,3,6,8-tetraone, ntda, unii-l56xqd1v6y, 2,7-dioxapyrene-1,3,6,8-tetrone, naphthalene-1,8:4,5-tetracarboxylic dianhydride, 1,8:4,5-naphthalenetetracarboxylic dianhydride, naphthalene-1,4,5,8-tetracarboxylic dianhydride
PubChem CID 6678
SMILES C1=CC2=C3C(=CC=C4C3=C1C(=O)OC4=O)C(=O)OC2=O

Specifications

Melting Point >300°C
Quantity 25 g
Beilstein 272788
Sensitivity Moisture sensitive
Formula Weight 268.18
Percent Purity 97%
Chemical Name or Material Naphthalene-1,4,5,8-tetracarboxylic acid dianhydride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.