missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Naphthalene-2,6-dicarboxylic acid, 98+%, Thermo Scientific Chemicals
£22.70 - £269.00
Chemical Identifiers
CAS | 1141-38-4 |
---|---|
Molecular Formula | C12H8O4 |
Molecular Weight (g/mol) | 216.192 |
MDL Number | MFCD00004105 |
InChI Key | RXOHFPCZGPKIRD-UHFFFAOYSA-N |
Synonym | 2,6-naphthalenedicarboxylic acid, 2,6-naphthalic acid, 2,6-dicarboxynaphthalene, unii-k3c4dyz29o, 2,6-naphthalene dicarboxylic acid, k3c4dyz29o, dsstox_cid_9211, acmc-2099kj, d06kmp, dsstox_rid_78711 |
PubChem CID | 14357 |
ChEBI | CHEBI:44460 |
IUPAC Name | naphthalene-2,6-dicarboxylic acid |
SMILES | C1=CC2=C(C=CC(=C2)C(=O)O)C=C1C(=O)O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11462898
|
Thermo Scientific Alfa Aesar
A16868.06 |
5 g |
£22.70
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11472898
|
Thermo Scientific Alfa Aesar
A16868.14 |
25 g |
£91.40
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11482898
|
Thermo Scientific Alfa Aesar
A16868.22 |
100 g |
£269.00
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1141-38-4 | |
216.192 | |
RXOHFPCZGPKIRD-UHFFFAOYSA-N | |
14357 | |
naphthalene-2,6-dicarboxylic acid |
C12H8O4 | |
MFCD00004105 | |
2,6-naphthalenedicarboxylic acid, 2,6-naphthalic acid, 2,6-dicarboxynaphthalene, unii-k3c4dyz29o, 2,6-naphthalene dicarboxylic acid, k3c4dyz29o, dsstox_cid_9211, acmc-2099kj, d06kmp, dsstox_rid_78711 | |
CHEBI:44460 | |
C1=CC2=C(C=CC(=C2)C(=O)O)C=C1C(=O)O |
Specifications
1141-38-4 | |
Odorless | |
MFCD00004105 | |
2,6-naphthalenedicarboxylic acid, 2,6-naphthalic acid, 2,6-dicarboxynaphthalene, unii-k3c4dyz29o, 2,6-naphthalene dicarboxylic acid, k3c4dyz29o, dsstox_cid_9211, acmc-2099kj, d06kmp, dsstox_rid_78711 | |
C1=CC2=C(C=CC(=C2)C(=O)O)C=C1C(=O)O | |
216.192 | |
CHEBI:44460 | |
≥98% |
>300°C | |
C12H8O4 | |
2051257 | |
RXOHFPCZGPKIRD-UHFFFAOYSA-N | |
naphthalene-2,6-dicarboxylic acid | |
14357 | |
216.19 | |
Naphthalene-2,6-dicarboxylic acid |
Safety and Handling
missing translation for 'einecsNumber' : 214-527-0
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only