missing translation for 'onlineSavingsMsg'
Learn More

Naphthalene, 99+%, scintillation grade

Product Code. 10124390 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
250 g
1 kg
Packaging:
Glass Bottle
Unit Size:
1kg
250g
5g
This item is not returnable. View return policy

Product Code. 10124390

Brand: Thermo Scientific Acros 180200010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 91-20-3 | C10H8 | 128.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For fluorescence/luminescence spectroscopy
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 91-20-3
Molecular Formula C10H8
Molecular Weight (g/mol) 128.17
MDL Number MFCD00001742
InChI Key UFWIBTONFRDIAS-UHFFFAOYSA-N
PubChem CID 931
ChEBI CHEBI:16482
IUPAC Name naphthalene
SMILES C1=CC2=CC=CC=C2C=C1

Specifications

Boiling Point 218.0°C
Melting Point 79.0°C to 82.0°C
Physical Form Powder
CAS Min % 99.0
CAS Max % 100.0
Density 0.9900g/mL
Color Beige to Red/Salmon
Quantity 1 kg
Flash Point 78°C
Assay Percent Range 99+%
Infrared Spectrum Authentic
Linear Formula C10H8
Beilstein 05, IV, 1640
Merck Index 15, 6455
Packaging Glass Bottle
Solubility Information Solubility in water: 30 mg/l (25°C). Other solubilities: soluble in ethanol,, very soluble in acetone, chloroform, ether,, benzene, toluene, carbon disulfide
Formula Weight 128.17
Percent Purity ≥99%
Specific Gravity 0.99
Chemical Name or Material Naphthalene, 99+%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.