missing translation for 'onlineSavingsMsg'
Learn More
Naphthalene, 99%, Thermo Scientific Chemicals Product Code.: 10153910

Naphthalene, 99%, Thermo Scientific Chemicals

Product Code. 10153910
250 g, Plastic bottle
Click to view available options
Quantity:
250 g
1 kg
2.5 kg
5 kg
Packaging:
Plastic bottle
Plastic drum
This item is not returnable. View return policy

Product Code. 10153910

Brand: Thermo Scientific Acros 180902500

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 91-20-3
Molecular Formula C10H8
Molecular Weight (g/mol) 128.17
MDL Number MFCD00001742
InChI Key UFWIBTONFRDIAS-UHFFFAOYSA-N
Synonym naphthalin, naphthene, camphor tar, tar camphor, white tar, albocarbon, naphthaline, moth flakes, moth balls, naphtalene
PubChem CID 931
ChEBI CHEBI:16482
IUPAC Name naphthalene
SMILES C1=CC2=CC=CC=C2C=C1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 79.0°C to 82.0°C
Color White
Density 0.9900g/mL
Boiling Point 218.0°C
Flash Point 78°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Plastic bottle
Linear Formula C10H8
Quantity 250 g
Beilstein 05,IV,164
Merck Index 15,6455
Specific Gravity 0.99
Solubility Information Solubility in water: 30mg/l (25°C). Other solubilities: soluble in ethanol,,very soluble in acetone,chloroform,ether,,benzene,toluene,carbon disulfide
Formula Weight 128.17
Percent Purity 99%
Physical Form Crystalline Flakes, Crystals, Tablets, Balls or Crystalline Powder
Chemical Name or Material Naphthalene
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0465623 Certificate TypeCertificate of Analysis Date22/08/2024 Catalog Number180902500,180900025,180900010,180900050
Lot NumberA0461639 Certificate TypeCertificate of Analysis Date05/04/2024 Catalog Number180902500,180900025,180900010,180900050
Lot NumberA0461638 Certificate TypeCertificate of Analysis Date05/04/2024 Catalog Number180902500,180900025,180900010,180900050
Lot NumberA0460958 Certificate TypeCertificate of Analysis Date12/03/2024 Catalog Number180902500,180900025,180900010,180900050
Lot NumberA0460959 Certificate TypeCertificate of Analysis Date11/03/2024 Catalog Number180902500,180900025,180900010,180900050
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Naphthalene
Signal Word
  • Warning
Hazard Category
  • Flammable solid Category 2
  • Acute toxicity Category 4
  • Carcinogenicity Category 2
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
  • H228-Flammable solid.
  • H302-Harmful if swallowed.
  • H351-Suspected of causing cancer.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P264-Wash thoroughly after handling.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P273-Avoid release to the environment.
  • Multiple Manufacturers
    Shop now and save up to 97% on top-quality products for your lab needs.
    Promo Code: 25908

    Expires2025-03-31

    Days

    0

    Hours

    11

    Minutes

    15

    Seconds

    11

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Naphthalene, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.