missing translation for 'onlineSavingsMsg'
Learn More

Naphthalene, 99%

Product Code. 10287023 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
250 g
1 kg
2.5 kg
5 kg
Packaging:
Plastic bottle
Plastic drum
Unit Size:
1kg
2.5kg
250g
5kg
This item is not returnable. View return policy

Product Code. 10287023

Brand: Thermo Scientific Acros 180900025

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 91-20-3 | C10H8 | 128.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 91-20-3
Molecular Formula C10H8
Molecular Weight (g/mol) 128.17
MDL Number MFCD00001742
InChI Key UFWIBTONFRDIAS-UHFFFAOYSA-N
Synonym naphthalin, naphthene, camphor tar, tar camphor, white tar, albocarbon, naphthaline, moth flakes, moth balls, naphtalene
PubChem CID 931
ChEBI CHEBI:16482
IUPAC Name naphthalene
SMILES C1=CC2=CC=CC=C2C=C1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 79.0°C to 82.0°C
Color White
Density 0.9900g/mL
Boiling Point 218.0°C
Flash Point 78°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Plastic bottle
Linear Formula C10H8
Quantity 2.5 kg
Beilstein 05,IV,164
Merck Index 15,6455
Specific Gravity 0.99
Solubility Information Solubility in water: 30mg/l (25°C). Other solubilities: soluble in ethanol,,very soluble in acetone,chloroform,ether,,benzene,toluene,carbon disulfide
Formula Weight 128.17
Percent Purity 99%
Physical Form Crystalline Flakes, Crystals, Tablets, Balls or Crystalline Powder
Chemical Name or Material Naphthalene
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.