Learn More
Naphthalene-d8, for NMR, +98% atom D, Thermo Scientific Chemicals
£56.68 - £215.07
Chemical Identifiers
CAS | 1146-65-2 |
---|---|
Molecular Formula | C10H8 |
Molecular Weight (g/mol) | 136.22 |
MDL Number | MFCD00001743 |
InChI Key | UFWIBTONFRDIAS-PGRXLJNUSA-N |
Synonym | naphthalene-d8, 2h8 naphthalene, perdeuteronaphthalene, naphthalene d8, perdeuterated naphthalene, 2 h? naphthalene, naphthalene-d8, analytical standard, naphthalene-d8 >98.0 atom % d, naphthalene d8 10 microg/ml in cyclohexane |
PubChem CID | 92148 |
IUPAC Name | 1,2,3,4,5,6,7,8-octadeuterionaphthalene |
SMILES | [2H]C1=C([2H])C2=C([2H])C([2H])=C([2H])C([2H])=C2C([2H])=C1[2H] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10605052
|
Thermo Scientific Acros
174960010 |
1 g | Glass Bottle |
£56.68
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10647504
|
Thermo Scientific Acros
174960050 |
5 g | Glass Bottle |
£215.07
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For NMRChemical Identifiers
1146-65-2 | |
136.22 | |
UFWIBTONFRDIAS-PGRXLJNUSA-N | |
92148 | |
[2H]C1=C([2H])C2=C([2H])C([2H])=C([2H])C([2H])=C2C([2H])=C1[2H] |
C10H8 | |
MFCD00001743 | |
naphthalene-d8, 2h8 naphthalene, perdeuteronaphthalene, naphthalene d8, perdeuterated naphthalene, 2 h? naphthalene, naphthalene-d8, analytical standard, naphthalene-d8 >98.0 atom % d, naphthalene d8 10 microg/ml in cyclohexane | |
1,2,3,4,5,6,7,8-octadeuterionaphthalene |
Specifications
Crystals or Crystalline Powder | |
98.0% min. | |
1146-65-2 | |
C10H8 | |
MFCD00001743 | |
UFWIBTONFRDIAS-PGRXLJNUSA-N | |
1,2,3,4,5,6,7,8-octadeuterionaphthalene | |
92148 | |
NMR |
White | |
78°C | |
81.0°C to 83.0°C | |
C10D8 | |
naphthalene-d8, 2h8 naphthalene, perdeuteronaphthalene, naphthalene d8, perdeuterated naphthalene, 2 h? naphthalene, naphthalene-d8, analytical standard, naphthalene-d8 >98.0 atom % d, naphthalene d8 10 microg/ml in cyclohexane | |
[2H]C1=C([2H])C2=C([2H])C([2H])=C([2H])C([2H])=C2C([2H])=C1[2H] | |
136.22 | |
136.22 | |
Naphthalene-d8, For NMR |
Safety and Handling
GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Flammable solid.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep container tightly closed.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
- Naphthalene-d8
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 1
- Carcinogenicity Category 2
- Flammable solid Category 2
Hazard Statement
- H228-Flammable solid.
- H302-Harmful if swallowed.
- H351-Suspected of causing cancer.
- H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
- P201-Obtain special instructions before use.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P370+P378-In case of fire: Use to extinguish.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.