missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Naproxen sodium, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | 259.0°C to 262.0°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 98% |
Packaging | Glass bottle |
Quantity | 5 g |
Merck Index | 15,6499 |
Formula Weight | 252.24 |
Percent Purity | 98% |
Chemical Name or Material | Naproxen sodium |
Safety and Handling


Product Identifier
- Naproxen sodium
- Danger
- Acute toxicity Category 4
- Reproductive toxicity Category 1A
- H302-Harmful if swallowed.
- H360-May damage fertility or the unborn child.
- P201-Obtain special instructions before use.
- P264-Wash face, hands and anyexposed skin thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- MIXTURE LIST-Contains : Sodium(-)-2-(6-methoxy-2-naphthyl)propionate
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Naproxen sodium, 98%, Thermo Scientific Chemicals >
Spot an opportunity for improvement?Share a Content Correction