missing translation for 'onlineSavingsMsg'
Learn More

BRAND™ Narrow Neck Erlenmeyer Flask

Narrow Neck Erlenmeyer Flask features a beaded rim and graduation marks.

Brand:  BRAND™ 92763

334.40 GBP valid until 2024-09-17
Use promo code "21690" to get your promotional price.



Additional Details : Weight : 54.81000kg

 View more versions of this product

Product Code. 15262979

  • £380.00 / Pack of 10
Estimated Shipment: 11-10-2024
to see stock.
Add to basket

Description

Description

  • Borosilicate Glass 3.3
  • DIN ISO 1773
  • With beaded rim and graduation
Specifications

Specifications

2000 mL
Narrow
Beaded
Borosilicate Glass (3.3)
Yes
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers