missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Duran™ Narrow Neck Reagent Bottle
Click to view available options
Capacity: 10mL
Specifications
Product Type | Reagent Bottle |
Capacity (Metric) | 10 mL |
Color | Clear |
Material | Glass |
Quantity | 10Pack |
Certified | Yes |
Certifications/Compliance | DIN 12035/ISO 4796 |
Mouth | Narrow |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'