Learn More
Nasco Whirl-Pak™ Dry Cellulose Sponge Probe Write-On Bag
GREENER_CHOICE
Convenient sterile absorbent sponge sampler in a sterile Whirl-Pak bag.
Brand:  Nasco B01475WA
206.15 GBP valid until 2024-12-31
Use promo code "21745" to get your promotional price.
Product Code. 16891590
  • £233.00 / Pack of 50
Explore more special offers
Description
Description
  • 8 in. (20.3 cm) long bendable polypropylene handle allows maximum contact of the sponge with the sampling surface
  • Helps with the collection of hard-to-reach surfaces, around corners, and irregularly shaped areas.
  • Innovative twist-off head holds the cellulose sponge securely and is quick and easy to remove from the handle once sample collection is completed.
  • Sterilized write-on bag.
Specifications
Specifications
Dry Sponge Probe
8 in.
50/Pk.
Designed for surface sampling
20.3 cm