missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Screw Top Vial Racks

Product Code. p-4520877
Click to view available options
Holds:
Vials
Vials (100)
For Use With (Equipment):
2 mL Vials
4 mL Vials
Description:
Vial Rack
Vial Rack with Lid
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 10620526

Brand: Thermo Scientific™ C401527

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Efficiently hold and organize vials while using them or for storage. Save space by stacking the 50 or 100 plastic vial racks on top of each other.

Store vials efficiently with Thermo Scientific™ National 4mL Screw Top Vial Racks, stackable for even more space savings.

  • Racks feature alphanumeric indexing for easier vial identification
  • Polypropylene vial racks are resistant to most solvents

Ordering Information
50 and 100 vial capacity racks available.

TRUSTED_SUSTAINABILITY

Specifications

Product Type Vial Rack
Description Vial Rack with Lid
Color Blue
Holds Vials (100)
Material Polystyrene
Shape Square
Stackable Yes
For Use With (Equipment) 4 mL Vials

Limited warranty period:
Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter.

Disclaimer to internal research use restriction:
Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.