missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ National Assembled Standard Opening Vial, Cap and Septum Kits

Product Code. p-4526376
Click to view available options
Color:
Amber
Clear
Closure Type:
Phenolic
Polypropylene
Polypropylene, Flanged
Septum:
PTFE
PTFE/Silicone
PTFE/Silicone, Pre-slit
PTFE/Silicone/PTFE
Packaging Type:
Polybag
Storage Tray
Unit Size:
Pack of 100
This item is not returnable. View return policy

Product Code. 11911613

Brand: Thermo Scientific™ undefined

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Thermo Scientific™ National Assembled Standard Opening Vial, Cap and Septum Kits provide convenience, efficiency and stackable storage options.

Thermo Scientific™ National Assembled Standard Opening Vial, Cap and Septum Kits provide convenience, efficiency and stackable storage options.

  • Packaged in convenient vial trays with clear covers or in economical polybags
  • Vial trays are stackable for easy storage
  • All caps are manufactured from virgin polypropylene unless noted

Includes: 

100 vials with pre-attached caps and septa

TRUSTED_SUSTAINABILITY

Specifications

Closure Color Black
Color (Seal) Red/White
Description Clear Glass; PP Black Cap; Red PTFE/White Silicone septum; in polybag
Color Clear
Material Glass, PTFE, Silicone
Volume (Metric) 2 mL
Septum PTFE/Silicone
Packaging Type Polybag
Closure Material Polypropylene
Closure Type Polypropylene
Closure Size 8-425
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.