missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Assembled Target™ 10-425 Caps and Septa

Minimize contamination from handling with convenient Thermo Scientific™ National Assembled Target 10-425 Caps and Septa.

£10.45 - £168.00

Specifications

Product Line National Assembled Target
Type Screw Cap
Closure Size 10 mm
For Use With (Equipment) 10-425 Screw Thread Vials
View More Specs

Products 2
Product Code Brand Color Septum Price Quantity & Availability  
Product Code Brand Color Septum Price Quantity & Availability  
12443658
View Documents
Thermo Scientific™
C4010-5A
Natural Integral Molded Polypropylene
£10.45
Pack of 100
Estimated Shipment: 31-01-2025
to see stock.
Add to basket
 
12483658
View Documents
Thermo Scientific™
C4010-60AY
Yellow Red PTFE/White Silicone This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

Minimize contamination from handling with convenient Thermo Scientific™ National Assembled Target 10-425 Caps and Septa.

  • Pre-assembled caps and septa are convenient and minimize contamination from handling
  • Storage cap has a solid top and pre-inserted PTFE disk liner
Specifications

Specifications

National Assembled Target
Screw Cap
10 mm
10-425 Screw Thread Vials
Videos
SDS
Documents

Documents

Product Certifications

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.