missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ National Preassembled Target DP™ Kits
Thermo Scientific™ National Preassembled Target DP Kits include 100 vials with pre-attached caps and septa.
Brand: Thermo Scientific™ C4000186W
This item is not returnable.
View return policy
Description
Thermo Scientific™ National Preassembled Target DP Kits include 100 vials with pre-attached caps and septa.
- Packaged in convenient vial trays with clear covers or in polybags

Specifications
Blue | |
Clear | |
2 mL | |
9-425 |
2mL; Clear Glass; w/ID, PTFE/Silicone/PTFE Septa; Blue cap | |
Glass | |
PTFE/Silicone/PTFE | |
Yes |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Thermo Scientific™ National Preassembled Target DP™ Kits > 2mL; Clear Glass; w/ID, PTFE/Silicone/PTFE Septa; Blue cap
Spot an opportunity for improvement?Share a Content Correction