Learn More
Thermo Scientific™ National Screw Thread Caps with Bonded Septa
Effectively eliminate septa push-through with Thermo Scientific™ National Screw Thread Caps with Bonded Septa.
Brand: Thermo Scientific™ C4015-69A
Code : NEW
Additional Details : Weight : 0.01000kg
Description
Effectively eliminate septa push-through with Thermo Scientific™ National Screw Thread Caps with Bonded Septa.
- Septa and polypropylene caps bonded together for an integral system
- Eliminates septa push-through
- Available with choice of GPI thread sizes and preslit septa for easier thin-gauge needle penetration
Specifications
Black; Bonded Preslit Red 0.005 in. PTFE/White Silicone Septa; GPI Thread Finish: 13-425 | |
Yes | |
National |
PTFE, Silicone, Polypropylene | |
Bonded Red PTFE/White Silicone, 0.005 in. | |
13-425 |