Learn More
Thermo Scientific™ National Target™ Snap-It™ Kits
Product Code. p-4526338
Click to view available options
Marking Spot:
No
Yes
Closure Color:
Clear
Color:
Clear
Septum:
PTFE/Red Rubber
PTFE/Silicone, Pre-slit
Unit Size:
Pack of 100
Product Code. 10281135
Brand: Thermo Scientific™ C401172W
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Thermo Scientific™ National Target Snap-It Convenience Kits provide vials and Snap-it caps with prefitted septa in storage trays.
Thermo Scientific™ National Target Snap-It Convenience Kits provide vials and Snap-it caps with prefitted septa in storage trays.
  • 2mL,12 × 32mm vials
  • Choice of vial and preassembled seals with septa
  • Packaged in two compartment benchtop storage trays
  • Include matched quantities of vials and Snap-It caps with prefitted septa
  • All items remain dust-free
TRUSTED_SUSTAINABILITY
Specifications
Closure Color Clear
Description Clear Glass I-D; Septum: PTFE/Red Rubber
Color Clear
Material PTFE, Rubber
Volume (Metric) 2 mL
Septum PTFE/Red Rubber
Closure Material PTFE, Rubber
Marking Spot Yes