missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ National Unassembled Certified™ Vial Kits

Thermo Scientific™ National Unassembled Certified Vial Kits provide extra assurance that time after time, the product upholds rigorous manufacturing standards.

Brand:  Thermo Scientific™ CERT4015-86

 View more versions of this product

Product Code. 11951553

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

Thermo Scientific™ National Unassembled Certified Vial Kits provide extra assurance that time after time, the product upholds rigorous manufacturing standards.

  • Fully lot tested for HPLC and GC analysis
  • Certificate of Conformance in every pack
  • Bonded caps to prevent septa push-through
  • Clear Type 1, 33 expansion borosilicate glass or amber Type 2, 51 expansion borosilicate glass
  • Available with or without ID patch
  • Fixed micro vials or inserts for limited-volume sampling
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Certified 4mL Amber Glass Screw-top Vial Kit, 13-425 Screw Thread
PTFE, Silicone
PTFE/Silicone
Screw Cap
Amber
4 mL
Bonded PTFE, Silicone
13-425
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers