Learn More
Thermo Scientific™ National Unassembled Standard Opening Vial, Cap and Septum Convenience Kits
Keep all items handy and dust-free with Thermo Scientific™ National Unassembled Vial, Cap and Septum Convenience Kits.
Brand: Thermo Scientific™ C4013-16
This item is not returnable.
View return policy
Description
Keep all items handy and dust-free with Thermo Scientific™ National Unassembled Vial, Cap and Septum Convenience Kits.
- Two-compartment, see-through benchtop storage tray
- Caps with septa are not attached to vials (packaged in plastic bags)
Includes:
100 vials and 100 caps with pre-assembled septa
Specifications
Black | |
Clear | |
Red PTFE/White Silicone/Red PTFE |
Vials: Clear Glass; Closure: Black; Septa: Red PTFE/White Silicone/Red PTFE | |
PTFE, Silicone |