Learn More
Thermo Scientific™ National Unassembled Standard Opening Vial, Cap and Septum Convenience Kits
Keep all items handy and dust-free with Thermo Scientific™ National Unassembled Vial, Cap and Septum Convenience Kits.
£36.85 - £62.45
Product Code | Brand | Description | Septum | Color | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Septum | Color | Price | Quantity & Availability | |||
12460108
|
Thermo Scientific™
C4013-16 |
Vials: Clear Glass; Closure: Black; Septa: Red PTFE/White Silicone/Red PTFE | Red PTFE/White Silicone/Red PTFE | Clear | |||||
12450128
|
Thermo Scientific™
C4013-492W |
Vials: Clear I-D; Closure: Black Phenolic; Septa: Red PTFE/White Silicone | Red PTFE/White Silicone | Clear | |||||
12400168
|
Thermo Scientific™
C4013-S15 |
Vials: Clear, Silanized; Closure: Black; Septa: Red PTFE/White Silicone | Red PTFE/White Silicone | Clear | |||||
Description
Keep all items handy and dust-free with Thermo Scientific™ National Unassembled Vial, Cap and Septum Convenience Kits.
- Two-compartment, see-through benchtop storage tray
- Caps with septa are not attached to vials (packaged in plastic bags)
Includes:
100 vials and 100 caps with pre-assembled septa
Specifications
Black |