missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NativePAGE™ 3 to 12%, Bis-Tris, 1.0 mm, Mini Protein Gels

The Invitrogen NativePAGE Bis-Tris Gel System is a precast polyacrylamide mini-gel system that provides sensitive, high-resolution analysis of native proteins and protein complexes for molecular mass estimations, and assessment of purity.

£180.00

Specifications

For Use With (Equipment) Mini Gel Tank, XCell SureLock Mini-Cell
Gel Percentage 3 to 12%
Gel Size Mini
Gel Thickness 1.0 mm
Gel Type Bis-Tris
View More Specs

Products 2
Product Code Brand Wells Price Quantity & Availability  
Product Code Brand Wells Price Quantity & Availability  
10064012
View Documents Promotion Details
Invitrogen™
BN1001BOX
10-well
£180.00
Each
In Stock
to see stock.
Add to basket
 
10024112
View Documents Promotion Details
Invitrogen™
BN1003BOX
15-well
£180.00
Each
In Stock
to see stock.
Edge
Add to basket
 
Description

Description

The Invitrogen NativePAGE Bis-Tris Gel System is a precast polyacrylamide mini-gel system that provides sensitive, high-resolution analysis of native proteins and protein complexes for molecular mass estimations, and assessment of purity. It is based on the blue native polyacrylamide gel electrophoresis (BN PAGE) technique developed by Schägger and von Jagow, which overcomes the limitations of traditional native electrophoresis by providing a near-neutral operating pH and detergent compatibility.

Features of the NativePAGE Bis-Tris Gel System include:
Wide molecular weight resolving range, from 15 – 10,000 kDa
Neutral-pH separation, which better preserves the native state of protein complexes
Resolution of all proteins in the gel regardless of their isoelectric point (pI)
Ability to analyze membrane-protein complexes in their native conformations
Ability to obtain better resolution than with traditional tris-glycine native electrophoresis

How NativePAGE Bis-Tris Gels work
In SDS-PAGE, SDS functions as a charge-shift molecule that denatures proteins by conferring on them a net negative charge and enables proteins to migrate towards the anode. BN PAGE uses Coomassie G-250 as the charge-shift molecule, which binds to proteins and confers a net negative charge while maintaining the proteins in their nondenatured, native state. The near-neutral pH of the NativePAGE Bis-Tris Gel System provides maximum stability of both the proteins and the gel matrix, resulting in a highly sensitive method for analysis of native membrane-protein complexes and offering superior band resolution over traditional Tris-glycine gel systems.

For transferring proteins to a membrane, we recommend using NuPAGE Transfer Buffers for traditional wet transfer and PVDF membranes. The NuPAGE transfer buffer maintains the neutral pH environment established during electrophoresis. Nitrocellulose membranes are not compatible for blotting since the membrane will bind the Coomassie G-250 dye very tightly. Alternatively, Rapid semi-dry transfer can be done using the Invitrogen Power Blotter (PB0013) or rapid dry transfer using the iBlot 2 Gel Transfer Device (IB21001) using PVDF membranes.

Specifications

Specifications

Mini Gel Tank, XCell SureLock Mini-Cell
Mini
Bis-Tris
30 to 10,000 kDa
Molecular Weight
10 Gels/Box
Store at 2°C to 8°C. Do not freeze.
8 cm
3 to 12%
1.0 mm
Native
Native
NativePAGE™
Wet Ice
8 cm
Videos
Safety and Handling

Safety and Handling

missing translation for 'shelfLife' : 12 Months

SDS
Documents

Documents

Product Certifications

For Research Use Only. Not for use in diagnostic procedures.