Learn More
Fisherbrand™ Bunsen Safety Burner for LP Gas
Supplied with a flame failure device that shuts off the gas supply to the burner within approximately 20 seconds of the flame being extinguished.
Brand:  Fisherbrand™ 4006/2TC
Product Code. 12862144
  • £66.60 / Each
Estimated Shipment: 15-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Safety features all contained within the burner
  • For use with natural gas/methane
  • 12.7mm diameter opening
Specifications
Specifications
Natural Gas, Methane
1 Ea.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Fisherbrand™ Bunsen Safety Burner for LP Gas > Gas Type: Natural/Methane