missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ark Plas Products™ Natural Polypropylene Commercial Grade Threaded Straight Connectors
Use these fittings for a wide range of applications. Ark Plas Products™ Natural Polypropylene Commercial Grade Threaded Straight Connectors are designed as a low cost alternative with high end performance.
Marca: Ark Plas Products™ CGA56-PR0
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
- Economic and reliable
- Up to 125 psi without clamp
- Contain recycled runner material
- Available in Natural Nylon or Natural Polypropylene
Especificaciones
Threaded Straight Connector | |
4.8 mm | |
25 Pack | |
1/4-18 UNF Male Thread | |
Natural |
Autoclavable | |
Polypropylene | |
3/16 in. Barb | |
Straight |