Learn More
Ark Plas Products™ Natural Polypropylene Lock Ring
Use these luers with a wide variety of medical and laboratory instruments. Ark Plas Products™ Natural Polypropylene Lock Rings feature conical tapers molded to ISO 594-1 and ISO 594-2 Standards for 6% luer tapers to ensure compatibility with all other luers molded to the same standard.
Brand: Ark Plas Products™ LEX66-PP0
This item is not returnable.
View return policy
Description
- Natural polypropylene luers
- Feature barbs designed with no parting line for reliable, leak-free use with a broad variety of medical and laboratory instruments, including syringetips and needles or stopcocks and needles
- Molded in virgin, lot-traceable material
- Ark-Plas Luer fittings include a large selection of both male and female luers available in locking or slip tip variances, as well as locking rings, bulkhead luers for panel mounting, threaded luers, adapters and plugs
Specifications
Lock Ring | |
Polypropylene | |
25 Pack | |
Natural |