missing translation for 'onlineSavingsMsg'
Learn More

Saint Gobain Life Sciences Natural Rubber Folding Skirt Stoppers

Use these stoppers for extra protection of tube ends during transport, handling and other operations during which tubes require both internal and end protection. Saint Gobain Life Sciences™ Natural Rubber Folding Skirt Stoppers provide an excellent seal.

Brand:  Saint Gobain Life Sciences 407010-50

 View more versions of this product

Product Code. 11774588

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Stoppers are flexible and provide a complete seal and good adhesion
  • Grey-blue grade: made from natural rubber
  • Temperature resistance from -35 to +70°C (120°C in moist heat)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

10.4 to 13.4mm
50/Pk.
Stopper
8.5mmH/13mmH
Natural Rubber
10.2D
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers