missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Natural Rubber One Hole Stoppers
Used for laboratory purposes.
£5.05 - £70.70
Specifications
Color | Red |
---|---|
Material | Natural Rubber |
For Use With (Application) | For laboratory use |
Hardness | 46/55 IRHD |
Product Code | Brand | Diameter (Metric) Bottom | Diameter (Metric) Top | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) Bottom | Diameter (Metric) Top | Price | Quantity & Availability | |||||
11577592
|
Fisherbrand™
RB010/1H |
10 mm | 12.5 mm |
£6.10
Pack of 20 |
Estimated Shipment: 11-04-2025
Log in to see stock. |
|||||
11587592
|
Fisherbrand™
RB011/1H |
11 mm | 14 mm |
£8.10
Pack of 20 |
Estimated Shipment: 15-04-2025
Log in to see stock. |
|||||
11517602
|
Fisherbrand™
RB017/1H |
17 mm | 20.3 mm |
£35.72
Pack of 20 |
In Stock
Log in to see stock. |
|||||
11527602
|
Fisherbrand™
RB019/1H |
19 mm | 22.5 mm |
£27.17
Pack of 20 |
Estimated Shipment: 22-04-2025
Log in to see stock. |
|||||
11507602
|
Fisherbrand™
RB015/1H |
15 mm | 18 mm |
£17.20
Pack of 20 |
In Stock
Log in to see stock. ![]() |
|||||
11567622
|
Fisherbrand™
RB105/1H |
105 mm | 116.5 mm |
£37.40
Each |
Estimated Shipment: 16-04-2025
Log in to see stock. |
|||||
11547602
|
Fisherbrand™
RB023/1H |
23 mm | 26.5 mm |
£56.46
Pack of 20 |
Estimated Shipment: 22-04-2025
Log in to see stock. |
|||||
11537602
|
Fisherbrand™
RB021/1H |
21 mm | 24.5 mm |
£45.78
Pack of 20 |
In Stock
Log in to see stock. ![]() |
|||||
11557602
|
Fisherbrand™
RB025/1H |
25 mm | 28.5 mm |
£36.76
Pack of 20 |
Estimated Shipment: 29-04-2025
Log in to see stock. |
|||||
11567602
|
Fisherbrand™
RB027/1H |
27 mm | 31 mm |
£40.78
Pack of 10 |
In Stock
Log in to see stock. |
|||||
11577602
|
Fisherbrand™
RB029/1H |
29 mm | 33 mm |
£43.45
Pack of 10 |
Estimated Shipment: 22-04-2025
Log in to see stock. |
|||||
11587602
|
Fisherbrand™
RB031/1H |
31 mm | 36 mm |
£51.16
Pack of 10 |
Estimated Shipment: 22-04-2025
Log in to see stock. |
|||||
11547612
|
Fisherbrand™
RB043/1H |
43 mm | 49 mm |
£70.70
Pack of 5 |
In Stock
Log in to see stock. ![]() |
|||||
11597602
|
Fisherbrand™
RB033/1H |
33 mm | 38.5 mm |
£52.52
Pack of 10 |
In Stock
Log in to see stock. ![]() |
|||||
11567612
|
Fisherbrand™
RB049/1H |
49 mm | 55 mm |
£26.26
Pack of 5 |
In Stock
Log in to see stock. |
|||||
11527612
|
Fisherbrand™
RB039/1H |
39 mm | 44.5 mm |
£57.55
Pack of 10 |
Estimated Shipment: 15-04-2025
Log in to see stock. |
|||||
11587612
|
Fisherbrand™
RB057/1H |
57 mm | 64 mm |
£57.55
Pack of 2 |
In Stock
Log in to see stock. ![]() |
|||||
11527592
|
Fisherbrand™
RB005/1H |
5 mm | 7 mm |
£11.15
Pack of 50 |
Estimated Shipment: 20-05-2025
Log in to see stock. |
|||||
11597612
|
Fisherbrand™
RB062/1H |
62 mm | 69 mm |
£47.27
Pack of 2 |
Estimated Shipment: 28-04-2025
Log in to see stock. |
|||||
11507622
|
Fisherbrand™
RB067/1H |
67 mm | 74 mm |
£68.53
Pack of 2 |
Estimated Shipment: 29-04-2025
Log in to see stock. |
|||||
11517622
|
Fisherbrand™
RB071/1H |
71 mm | 78 mm |
£28.39
Each |
Estimated Shipment: 15-04-2025
Log in to see stock. |
|||||
11527622
|
Fisherbrand™
RB075/1H |
75 mm | 85 mm |
£31.51
Each |
Estimated Shipment: 15-04-2025
Log in to see stock. |
|||||
11547592
|
Fisherbrand™
RB007/1H |
7 mm | 9 mm |
£5.23
Pack of 20 |
Estimated Shipment: 15-04-2025
Log in to see stock. |
|||||
11557592
|
Fisherbrand™
RB008/1H |
8 mm | 10.5 mm |
£5.23
Pack of 20 |
Estimated Shipment: 01-05-2025
Log in to see stock. |
|||||
11547622
|
Fisherbrand™
RB092/1H |
92 mm | 102 mm |
£30.47
Each |
Estimated Shipment: 29-05-2025
Log in to see stock. |
|||||
10410893
|
Fisherbrand™
RB009/1H |
9 mm | 11.5 mm | N/A | ||||||
11567592
|
Fisherbrand™
RB009/1H |
9 mm | 11.5 mm |
£5.05
Pack of 20 |
Estimated Shipment: 29-05-2025
Log in to see stock. |
|||||
Description
- Made up of natural rubber
- Red in color
- Contains one hole
Spécification
Red | |
Natural Rubber | |
For laboratory use | |
46/55 IRHD |
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Fisherbrand™ Natural Rubber One Hole Stoppers