missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRAND™ Natural Rubber Pipet Filler
Manufactured from 100% natural rubber. BRAND™ Natural Rubber Pipet Filler eliminates mouth pipetting and handling of hazardous material. Available in universal, standard, or flip model, with 2 or 3-valve options.
£17.60 - £38.96
Product Code | Brand | Volume (Metric) | Model | Includes | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Model | Includes | Price | Quantity & Availability | |||
10363341
|
BRAND™
25315 |
100 mL | Universal | 3 valves |
£38.96
Each |
In Stock
Log in to see stock. |
|||
10221601
|
BRAND™
25300 |
10 mL | Standard | 3 valves |
£20.38
Each |
In Stock
Log in to see stock. |
|||
10110671
|
BRAND™
25400 |
100 mL | Flip | 2 valves |
£17.60
Each |
Estimated Shipment: 26-07-2024
Log in to see stock. |
|||
Description
- Manufactured from 100% natural rubber.
- Eliminates mouth pipetting and handling of hazardous material.
- Available in universal, standard, or flip model, with 2 or 3-valve options
- 2-valve flip model enables one-handed operation.
Specifications
Natural Rubber | |
1 Each |