missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Solid Natural Rubber Stoppers
Used for laboratory purposes.
£11.15 - £63.65
Specifications
Color | Red |
---|---|
Material | Natural Rubber |
For Use With (Application) | For laboratory use |
Hardness | 46/55 IRHD |
Product Code | Brand | Diameter (Metric) Bottom | Diameter (Metric) Top | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) Bottom | Diameter (Metric) Top | Price | Quantity & Availability | |||||
11517592
|
Fisherbrand™
RB105/S |
105 mm | 116.5 mm |
£37.40
Each |
Estimated Shipment: 15-04-2025
Log in to see stock. |
|||||
11522922
|
Fisherbrand™
RB011/S |
11 mm | 14 mm |
£21.01
Pack of 20 |
Estimated Shipment: 13-05-2025
Log in to see stock. |
|||||
11552922
|
Fisherbrand™
RB017/S |
17 mm | 20.3 mm |
£33.35
Pack of 20 |
In Stock
Log in to see stock. ![]() |
|||||
11542932
|
Fisherbrand™
RB035/S |
35 mm | 40.5 mm |
£52.50
Pack of 10 |
Estimated Shipment: 15-04-2025
Log in to see stock. |
|||||
11522942
|
Fisherbrand™
RB057/S |
57 mm | 64 mm |
£33.64
Pack of 2 |
Estimated Shipment: 15-04-2025
Log in to see stock. |
|||||
11532942
|
Fisherbrand™
RB062/S |
62 mm | 69 mm |
£63.65
Pack of 2 |
Estimated Shipment: 15-04-2025
Log in to see stock. |
|||||
11542942
|
Fisherbrand™
RB067/S |
67 mm | 74 mm |
£26.26
Pack of 2 |
Estimated Shipment: 15-04-2025
Log in to see stock. |
|||||
11572912
|
Fisherbrand™
RB006/S |
6 mm | 8 mm |
£11.15
Pack of 50 |
Estimated Shipment: 15-04-2025
Log in to see stock. |
|||||
11582922
|
Fisherbrand™
RB023/S |
23 mm | 26.5 mm |
£55.26
Pack of 20 |
In Stock
Log in to see stock. ![]() |
|||||
10192031
|
Fisherbrand™
RB023/S |
23 mm | 26.5 mm | N/A | ||||||
10411083
|
Fisherbrand™
RB025/S |
25 mm | 28.5 mm | N/A | ||||||
11592922
|
Fisherbrand™
RB025/S |
25 mm | 28.5 mm |
£34.35
Pack of 20 |
Estimated Shipment: 15-04-2025
Log in to see stock. |
|||||
Description
- Made up of natural rubber
- Red in color
- Solid stoppers with no holes
Specifications
Red | |
Natural Rubber | |
For laboratory use | |
46/55 IRHD |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Fisherbrand™ Solid Natural Rubber Stoppers