Learn More
Abeba™ Nature 4085 Shoes
Leather covered cork footbed is ESD dissipative.
Brand: Abeba™ 4085/38
Additional Details : Weight : 0.80000kg
Description
- CE EN ISO 20347:2012 OB A, FO SRC
- Smooth leather
- Black
- Slip resistance special rubber outsole sole
- Leather coated cork footbed
- Sanitized treated
- ESD dissipative according to DIN EN 61340
- 2 adjustable instep straps
- velcro fastening
Specifications
CE EN ISO 20347:2012 OB A, FO SRC | |
Cork | |
38 | |
Rubber |
Black | |
Leather | |
Slip-resistant | |
4085 |