Learn More
Abeba™ Nature 4085 Shoes
Leather covered cork footbed is ESD dissipative.
Brand:  Abeba™ 4085/35
Additional Details : Weight : 0.80000kg
 View more versions of this product
Product Code. 15681438
  • £65.99 / 1 pair
Estimated Shipment: 15-08-2024
to see stock.
Add to basket
Description
Description
  • CE EN ISO 20347:2012 OB A, FO SRC
  • Smooth leather
  • Black
  • Slip resistance special rubber outsole sole
  • Leather coated cork footbed
  • Sanitized treated
  • ESD dissipative according to DIN EN 61340
  • 2 adjustable instep straps
  • velcro fastening
Specifications
Specifications
CE EN ISO 20347:2012 OB A, FO SRC
Cork
35
Rubber
Black
Leather
Slip-resistant
4085