missing translation for 'onlineSavingsMsg'
Learn More

NBD chloride, 98%, Thermo Scientific Chemicals

£23.32 - £141.71

Chemical Identifiers

CAS 10199-89-0
Molecular Formula C6H2ClN3O3
Molecular Weight (g/mol) 199.55
MDL Number MFCD00005808
InChI Key IGHBXJSNZCFXNK-UHFFFAOYSA-N
Synonym 4-Chloro-7-nitrobenzofurazan, 4-Chloro-7-nitrobenzo-2-oxa-1, 3-diazole
PubChem CID 25043
ChEBI CHEBI:78878
IUPAC Name 4-chloro-7-nitro-2,1,3-benzoxadiazole
SMILES C1=C(C2=NON=C2C(=C1)Cl)[N+](=O)[O-]
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10347880
View Documents
Thermo Scientific Acros
172390010
1 g Glass bottle
£23.32
1g
In Stock
to see stock.
Add to basket
 
10721451
View Documents Promotion Details
Thermo Scientific Acros
172390050
5 g Glass bottle
£43.62
5g
In Stock
to see stock.
Add to basket
 
10532924
View Documents
Thermo Scientific Acros
172390250
25 g Glass bottle
£141.71
25g
In Stock
to see stock.
Add to basket
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

10199-89-0
199.55
IGHBXJSNZCFXNK-UHFFFAOYSA-N
25043
4-chloro-7-nitro-2,1,3-benzoxadiazole
C6H2ClN3O3
MFCD00005808
4-Chloro-7-nitrobenzofurazan, 4-Chloro-7-nitrobenzo-2-oxa-1, 3-diazole
CHEBI:78878
C1=C(C2=NON=C2C(=C1)Cl)[N+](=O)[O-]

Specifications

96.0°C to 99.0°C
97.5
98%
MFCD00005808
Solubility in water: soluble.
Authentic
4-chloro-7-nitro-2,1,3-benzoxadiazole
25043
199.55
Brown to Yellow
NBD chloride
10199-89-0
100.0
C6H2ClN3O3
4-Chloro-7-nitrobenzofurazan, 4-Chloro-7-nitrobenzo-2-oxa-1, 3-diazole
IGHBXJSNZCFXNK-UHFFFAOYSA-N
C1=C(C2=NON=C2C(=C1)Cl)[N+](=O)[O-]
199.55
CHEBI:78878
98%
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

missing translation for 'einecsNumber' : 233-496-4

missing translation for 'rtecsNumber' : DF8002400

missing translation for 'tsca' : TSCA

Product Identifier
  • NBD chloride

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

Supplemental information
  • MIXTURE LIST-Contains : 4-chloro-7-nitro-2,1,3-benzoxadiazole

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only