missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ NBI 35965 hydrochloride
GREENER_CHOICE

Product Code. 18075422
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18075422

Brand: Tocris Bioscience™ 3100/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent and selective CRF1 antagonist

Potent and selective corticotropin-releasing factor receptor 1 (CRF1) antagonist (Ki values are 4 and > 10000 nM at CRF1 and CRF2 respectively). Inhibits cAMP accumulation and adrenocorticotropic hormone (ACTH) production in vitro (pIC50 values are 7.1 and 6.9 respectively) and attenuates CRF and ACTH production in vivo. Orally active and brain penetrant with anxiolytic activity.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 603151-83-3
Molecular Formula C21H23Cl3N4

Specifications

Quantity 10 mg
Formula Weight 437.79
Purity >98%
Molecular Weight (g/mol) 437.793
Chemical Name or Material NBI 35965 hydrochloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.