Learn More
NBN Mouse anti-Human, Unconjugated, Clone: 1E11E10, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 66980-1-IG-150UL
 View more versions of this product
Product Code. 16819325
  • £349.00 / 150µL
Explore more special offers
Description
Description
The NBN antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human NBN. This antibody recognizes human antigen. The NBN antibody has been validated for the following applications: IF, WB, ELISA analysis.
Specifications
Specifications
NBN
Monoclonal
1000 μg/mL
Mouse
Primary
Human
Western Blot, Immunofluorescence, ELISA
1E11E10
Unconjugated
150 μL
4683
IgG2a