Learn More
NBN Mouse anti-Human, Unconjugated, Clone: 1E11E10, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66980-1-IG-20UL
This item is not returnable.
View return policy
Description
The NBN antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human NBN. This antibody recognizes human antigen. The NBN antibody has been validated for the following applications: IF, WB, ELISA analysis.
Specifications
NBN | |
Monoclonal | |
1000 μg/mL | |
Mouse | |
Primary | |
Human |
Western Blot, Immunofluorescence, ELISA | |
1E11E10 | |
Unconjugated | |
20 μL | |
4683 | |
IgG2a |