missing translation for 'onlineSavingsMsg'
Learn More

NCAM2 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Brand:  Invitrogen PA556584

Product Code. 15764491

  • £427.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Immunogen sequence: NVPPAISMPQ KSFNATAERG EEMTFSCRAS GSPEPAISWF RNGKLIEENE KYILKGSNTE LTVRNIINSD GGPY Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 91%.

NCAM2 (neural cell adhesion molecule 2) is a cell adhesion molecule belonging to the immunoglobulin superfamily. It is differentially expressed in the developing and mature olfactory epithelium and is involved in the guidance of developing olfactory sensory neurons to the glomeruli of the olfactory bulb.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

NCAM2
Polyclonal
Unconjugated
NCAM2
fasciclin II; NCAM2; N-CAM-2; Ncam-2; NCAM21; neural cell adhesion molecule 2; Neural cell adhesion molecule RB-8; Ocam; OCAM-GPI; olfactory axon cell adhesion molecule; R4B12; R4B12 antigen; Rncam
Rabbit
Antigen affinity chromatography
RUO
17968, 4685
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry
0.1 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
O15394, O35136
NCAM2
Recombinant protein corresponding to Human NCAM2
100 μL
Primary
Human, Mouse
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Research Use Only