missing translation for 'onlineSavingsMsg'
Learn More

NCOA7 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

£135.00 - £349.00

Specifications

Antigen NCOA7
Concentration 0.37 mg/mL
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16802195
View Documents
Proteintech
23092-1-AP-20UL
20 μL
£135.00
20µL
Please to purchase this item. Need a web account? Register with us today!
16892185
View Documents
Proteintech
23092-1-AP-150UL
150 μL
£349.00
150µL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This antibody is a rabbit polyclonal antibody raised against the N-terminal of human NCOA7 protein.

NCOA7 enhances the transcriptional activities of several nuclear receptors. It is involved in the coactivation of different nuclear receptors, such as ESR1, THRB, PPARG and RARA.
Specifications

Specifications

NCOA7
Western Blot, Immunoprecipitation
Unconjugated
Rabbit
Human
Q8NI08
135112
NCOA7 Fusion Protein Ag19315
Primary
-20°C
NCOA7
0.37 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
ERAP140, ESNA1, Nbla00052, Nbla10993, NCOA7, nuclear receptor coactivator 7
NCOA7
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.