Learn More
Thermo Scientific™ NdeI (10 U/μL)
The NdeI restriction enzyme recognizes CA^TATG sites and cuts best at 37°C in O buffer (Isoschizomers: FauNDI).
£33.15 - £142.00
Specifications
Concentration 10 U/μL
View More Specs
Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10704491
View Documents Promotion Details
Thermo Scientific™
ER0581
500 U
£33.15
500 units
10160740
View Documents
Thermo Scientific™
ER0585
4000 Reactions
£142.00
4000 units
10818810
View Documents Promotion Details
Thermo Scientific™
ER0582
2500 U
£89.50
2500 units
Description
Description
Conventional restriction endonucleases are a large collection of high quality restriction enzymes, optimized to work in one of the buffers of the Five Buffer System. In addition, the universal Tango buffer is provided for convenience in double digestions. All of the enzymes exhibit 100% activity in the recommended buffer and reaction conditions. To ensure consistent performance, Thermo Scientific restriction enzyme reaction buffers contain premixed BSA, which enhances the stability of many enzymes and binds contaminants that may be present in DNA preparations.
Highlights
  • Superior quality – stringent quality control and industry leading manufacturing process
  • Convenient color-coded Five Buffer System
  • Includes universal Tango buffer for double-digestions
  • BSA premixed in reaction buffers
  • Wide selection of restriction endonuclease specificities
Specifications
Specifications
10 U/μL