missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NDEL1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17262-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene encodes a thiol-activated peptidase that is phosphorylated in M phase of the cell cycle. Phosphorylation regulates the cell cycle-dependent distribution of this protein, with a fraction of the protein bound strongly to centrosomes in interphase and localized to mitotic spindles in early M phase. Overall, this protein plays a role in nervous system development. Alternate transcriptional splice variants, encoding different isoforms, have been characterized.Specifications
NDEL1 | |
Polyclonal | |
Unconjugated | |
NDEL1 | |
EOPA, MITAP1, Mitosin associated protein 1, NDEL1, NUDEL, Protein Nudel | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
170845, 81565, 83431 | |
-20°C | |
Liquid |
Immunoprecipitation, Western Blot | |
0.43 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q78PB6, Q9ERR1, Q9GZM8 | |
Ndel1 | |
NDEL1 Fusion Protein Ag10975 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction