Learn More
NDEL1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 17262-1-AP-20UL
 View more versions of this product
Product Code. 16867124
  • £135.00 / 20µL
Explore more special offers
Description
Description
This gene encodes a thiol-activated peptidase that is phosphorylated in M phase of the cell cycle. Phosphorylation regulates the cell cycle-dependent distribution of this protein, with a fraction of the protein bound strongly to centrosomes in interphase and localized to mitotic spindles in early M phase. Overall, this protein plays a role in nervous system development. Alternate transcriptional splice variants, encoding different isoforms, have been characterized.
Specifications
Specifications
NDEL1
Polyclonal
Unconjugated
NDEL1
EOPA, MITAP1, Mitosin associated protein 1, NDEL1, NUDEL, Protein Nudel
Rabbit
Antigen Affinity Chromatography
RUO
170845, 81565, 83431
-20°C
Liquid
Immunoprecipitation, Western Blot
0.43 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q78PB6, Q9ERR1, Q9GZM8
Ndel1
NDEL1 Fusion Protein Ag10975
20 μL
Primary
Human, Rat, Mouse