missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ndelta-Benzyloxycarbonyl-Nalpha-Boc-D-ornithine, 98%, Thermo Scientific™
POA - £227.00
Chemical Identifiers
CAS | 16937-92-1 |
---|---|
Molecular Formula | C18H26N2O6 |
Molecular Weight (g/mol) | 366.41 |
MDL Number | MFCD00069970 |
InChI Key | QYYCZJUFHDLLOJ-UHFFFAOYNA-N |
Synonym | boc-d-orn z-oh, r-5-benzyloxy carbonyl amino-2-tert-butoxycarbonyl amino pentanoic acid, n-boc-n'-cbz-d-ornithine, 2r-5-benzyloxy carbonyl amino-2-tert-butoxycarbonyl amino pentanoic acid, ksc918o2r, n, a-boc-n, a-z-d-ornithine, a-z-n, a-boc-d-ornithine, nalpha-boc-ndelta-cbz-r-ornithine, boc-d-orn z-oh tlc, 2r-5-benzyloxycarbonylamino-2-tert-butoxycarbonylamino pentanoic acid |
PubChem CID | 7272030 |
SMILES | CC(C)(C)OC(=O)NC(CCCNC(=O)OCC1=CC=CC=C1)C(O)=O |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
16937-92-1 | |
366.41 | |
QYYCZJUFHDLLOJ-UHFFFAOYNA-N | |
7272030 |
C18H26N2O6 | |
MFCD00069970 | |
boc-d-orn z-oh, r-5-benzyloxy carbonyl amino-2-tert-butoxycarbonyl amino pentanoic acid, n-boc-n'-cbz-d-ornithine, 2r-5-benzyloxy carbonyl amino-2-tert-butoxycarbonyl amino pentanoic acid, ksc918o2r, n, a-boc-n, a-z-d-ornithine, a-z-n, a-boc-d-ornithine, nalpha-boc-ndelta-cbz-r-ornithine, boc-d-orn z-oh tlc, 2r-5-benzyloxycarbonylamino-2-tert-butoxycarbonylamino pentanoic acid | |
CC(C)(C)OC(=O)NC(CCCNC(=O)OCC1=CC=CC=C1)C(O)=O |
Specifications
16937-92-1 | |
MFCD00069970 | |
boc-d-orn z-oh, r-5-benzyloxy carbonyl amino-2-tert-butoxycarbonyl amino pentanoic acid, n-boc-n'-cbz-d-ornithine, 2r-5-benzyloxy carbonyl amino-2-tert-butoxycarbonyl amino pentanoic acid, ksc918o2r, n, a-boc-n, a-z-d-ornithine, a-z-n, a-boc-d-ornithine, nalpha-boc-ndelta-cbz-r-ornithine, boc-d-orn z-oh tlc, 2r-5-benzyloxycarbonylamino-2-tert-butoxycarbonylamino pentanoic acid | |
CC(C)(C)OC(=O)NC(CCCNC(=O)OCC1=CC=CC=C1)C(O)=O | |
366.41 | |
366.41 | |
Solid |
C18H26N2O6 | |
2400891 | |
QYYCZJUFHDLLOJ-UHFFFAOYNA-N | |
5-{[(benzyloxy)carbonyl]amino}-2-{[(tert-butoxy)carbonyl]amino}pentanoic acid | |
7272030 | |
98% | |
Ndelta-Benzyloxycarbonyl-Nalpha-Boc-D-ornithine |
Safety and Handling
missing translation for 'tsca' : No
missing translation for 'storageNote' : 2° to 8°C
Recommended Storage : Keep cold